Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purmorphamine, Tocris Bioscience™

Catalog No. p-4835957 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Smo receptor agonist

Purmorphamine is a smoothened (Smo) receptor agonist (EC50 ∼ 1 μM) and activates the Hedgehog pathway. Purmorphamine induces osteogenesis in mouse mesenchymal progenitor cells (C3H10T1/2). When combined with BMP-4, Purmorphamine can transdifferentiate pre-adipocytes (3T3-L1) and myoblasts (C2C12) into osteoblasts. Purmorphamine induces differentiation of multipotent mesenchymal progenitor cells into osteoblasts, and of spinal motor neurons from pluripotent human stem cells, and protects dopaminergic neurons in a mouse model of Parkinson's disease.

Chemical Identifiers

CAS 483367-10-8
Molecular Formula C31H32N6O2
Molecular Weight (g/mol) 520.637
InChI Key FYBHCRQFSFYWPY-UHFFFAOYSA-N
Synonym purmorphamine, 2-1-naphthoxy-6-4-morpholinoanilino-9-cyclohexylpurine, unii-pb12m2f8ky, shh signaling antagonist vi, pb12m2f8ky, 9-cyclohexyl-n-4-morpholin-4-yl phenyl-2-naphthalen-1-yloxy-9h-purin-6-amine, 9-cyclohexyl-n-4-4-morpholinyl phenyl-2-1-naphthalenyloxy-9h-purin-6-amine, 2-1-napthoxy-6-4-morpholinoanilino-9-cyclohexylpurine, 9-cyclohexyl-n-4-morpholin-4-ylphenyl-2-naphthalen-1-yloxypurin-6-amine
PubChem CID 5284329
ChEBI CHEBI:63053
IUPAC Name 9-cyclohexyl-N-(4-morpholin-4-ylphenyl)-2-naphthalen-1-yloxypurin-6-amine
SMILES C1CCC(CC1)N2C=NC3=C2N=C(N=C3NC4=CC=C(C=C4)N5CCOCC5)OC6=CC=CC7=CC=CC=C76

Specifications

Quantity 10 mg
Formula Weight 520.62
Chemical Name or Material Purmorphamine
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.