Learn More
Enzo Life Sciences Puromycin. 2HCl (250mg). CAS: 58-58-2
Supplier: Enzo Life Sciences BMLGR3120250
Inhibits protein synthesis. Inhibits translation by causing premature release of nascent polypeptide. Induces apoptosis. Alternative name: 3'-[[2-Amino-3-(4-methoxyphenyl)-1-oxopropyl]amino]-3'-deoxy-N,N-dimethyl-adenosine . 2HCl, 3'-(alpha-Amino-p-methoxy-hydrocinnamamido-3'-deoxy-N,N-dimethyl-adenosine . 2HCl. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.