Learn More
Enzo Life Sciences Puromycin . 2HCl, (50mg), CAS Number: 58-58-2
SDP
Supplier:  Enzo Life Sciences BMLGR3120050
Encompass_Preferred
Protein synthesis inhibitor. Inhibits protein synthesis. Inhibits translation by causing premature release of nascent polypeptide. Induces apoptosis. Alternative Name: 3'-[[2-Amino-3-(4-methoxyphenyl)-1-oxopropyl]amino]-3'-deoxy-N,N-dimethyl-adenosine . 2HCl, 3'-(alpha-Amino-p-methoxy-hydrocinnamamido-3'-deoxy-N,N-dimethyl-adenosine . 2HCl. Formula: C22H29N7O5 . 2HCl. MW: 544.4. CAS Number: 58-58-2. MI: 14: 7943. Purity: ≥98% (HPLC, TLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC9895494
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.