Learn More
Purus™ Cleanroom Tacky Mat: PROMO
Shop All Purus International Inc. ProductsSpecifications
Type
Purus Cleanroom Tacky Mat
Color
White
Material
LDPE
For Use With (Application)
For cleanroom purposes
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.