Learn More
ACROBiosystems ACROBiosystems Cynomolgus PVRIG Protein, Fc Tag
Recombinant Protein;100UG;Cynomolgus PVRIG, Fc Tag (PVG-C5259) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession # A0A2K5WVV8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.1 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-2, Mouse IgG2a Fc Tag (Cat. No. CD2-H5257) at 1 μg/mL (100 μL/well) can bind Cynomolgus PVRIG, Fc Tag (Cat. No. PVG-C5259) with a linear range of 0.5-8 ng/mL (QC tested).