Learn More
Beckman Coulter Biomek Tips P30XL Non-Sterile
Supplier:  Beckman Coulter A22287
Encompass
The AP384 P30XL non-sterile tips are designed to work with the Biomek 384-multichannel head. These tips offer an extended, narrow length of 43 mm, which allows pipetting from the bottom of deep labware to retrieve valuable samples. The tips also provide a larger transfer volume capacity of 50 µL, an additional 20 µL over the standard P30 tip. The AP384 P30XLs are 100% polypropylene, automation-grade and manufactured to ensure straight, leak-proof products free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. Case of 10 racks.
Catalog No. NC9281446
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.