Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid, 95%, Thermo Scientific™

Catalog No. AC449920010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 25940-35-6
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD01975194
InChI Key HNYVPKNVKSTVJO-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-3-carboxylic acid, pyrazolo 1,5-a pyrimidine-3-carboxylicacid, 3-carboxypyrazolo 1,5-a pyrimidine, 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylic acid, pubchem20936, acmc-1cbw6, 3-pyrazolo 1,5-a pyrimidinecarboxylic acid
PubChem CID 1910189
IUPAC Name pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
SMILES OC(=O)C1=C2N=CC=CN2N=C1

Specifications

Melting Point 275.0°C to 280.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.13
Percent Purity 95%
Chemical Name or Material Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.