Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREXPLUS™ Media/Solution Bottles

Catalog No. p-42940 Shop All Corning Products
Click to view available options
Capacity:
17 oz., 500 mL
3.3 oz., 100 mL
33.8 oz., 1000 mL
67.62 oz., 2000 mL
8.45 oz., 250 mL
Diameter (English):
2.2 in.
2.7 in.
3.3 in.
3.9 in.
5.3 in.
Graduations:
100 to 400 mL
100 to 900 mL
40 to 80 mL
400 to 1800 mL
50 to 200 mL
Height (English):
3.9 in.
5.4 in.
6.9 in.
8.8 in.
10.3 in.

Heavy-duty PYREXPLUS™ round bottles have a protective PVC coating to help prevent glass from shattering and reduces spills

Heavy duty bottle which can be used for storage as well as mixing and sampling. A protective PVC coating helps prevent glass from shattering and reduces spills. Autoclavable (121°C) and resistant to thermal shock. Bottle comes complete with autoclavable, one-piece, green colored, polypropylene plug seal cap with drip-free pouring ring. Glass bead indicates full capacity line. Teal enameled graduations and marking spot. Thread size: GL 45. Neck opening: 29 mm I.D.

Order Info

Various other caps fit this bottle and are available for separate purchase: Polypropylene colored caps in a choice of orange, green, purple, red, and light gray for ease of sorting and identification; Vented membrane cap with 0.22μm PTFE hydrophobic membrane; PBT caps with silicone septa for syringe sampling or introduction of reagents.

Specifications

Capacity 3.3 oz., 100 mL
Capacity (Metric) 100 mL
Capacity (English) 3.3 oz.
Coating Pyrexplus
Diameter (English) 2.2 in.
Material Borosilicate Glass
No. per Case 4/Cs.
No. per Pack 4/Pk.
Sterility Non-sterile
Closure Color Green
Graduations 40 to 80 mL
Mouth Wide
Shape Cylindrical
Height (Metric) 100 mm
Height (English) 3.9 in.
Closure Material Polypropylene
Closure Size GL 45
Diameter (Metric) 56 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.