Learn More
New England Biolabs, Inc. Q5 High-Fidelity 2X Master Mix – 100 reactions (50 µl vol)
SDP
Supplier:  New England Biolabs, Inc. M0492S
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
The Q5 High-Fidelity 2X Master Mix offers robust, high-fidelity performance in a convenient master mix format. The Q5 High- Fidelity 2X Master Mix features a high-fidelity, thermostable DNA polymerase with 3' to 5' exonuclease activity, fused to a processivity-enhancing Sso7d domain to support robust DNA amplification. With an error rate ~280-fold lower than that of Taq DNA Polymerase, Q5 High-Fidelity DNA Polymerase is ideal for cloning and can be used for long or difficult amplicons. The convenient master mix formulation is supplied at a 2X concentration. The mix contains dNTPs, Mg++ and a proprietary broad-use buffer requiring only the addition of primers and DNA template for robust amplification regardless of GC content. When used at the recommended 1X final concentration, the Q5 High-Fidelity Master Mix contains 2 mM Mg++. Q5 High-Fidelity DNA Polymerase is unlike typical, lower fidelity PCR enzymes.
Catalog No. NC0355745
missing translation for 'QuantityLeft'