Learn More
New England Biolabs, Inc. Q5 Site-Directed Mutagenesis Kit (Without Competent Cells) - 10 rxns
SDP
Supplier:  New England Biolabs, Inc. E0552S
Encompass
Q5 Site-Directed Mutagenesis Kit (Without Competent Cells) - 10 rxns. The Q5 Site-Directed Mutagenesis Kit (Without Competent Cells) enables rapid, site-specific mutagenesis of double-stranded plasmid DNA in less than 2 hours. The kit utilizes the robust Q5 Hot Start High-Fidelity DNA Polymerase along with custom mutagenic primers to create insertions, deletions and substitutions in a wide variety of plasmids. After PCR, the amplified material is added directly to a unique Kinase-Ligase-DpnI (KLD) enzyme mix for rapid (5 minutes), room temperature circularization and template removal. Transformation into high-efficiency chemically-competent E. coli, not supplied, ensures robust results with plasmids up to at least 20 kb in length.Help NEB go green. FREE FREIGHT on NEB orders greater than $350.00!
Catalog No. 50-607-647
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.