Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Clear Straight Sided Round Bottles with Green Thermoset F217 and PTFE Caps
Click to view available options
Capacity:
1 oz., 30mL
16 oz., 480 mL
2 oz., 60 mL
32 oz., 960 mL
4 oz., 120 mL
6 oz., 180 mL
8 oz., 240 mL
Height (Metric):
43 mm
49 mm
68 mm
79 mm
89 mm
96 mm
170 mm
Height (English):
1.69 in.
1.93 in.
2.68 in.
3.11 in.
3.5 in.
3.78 in.
6.69 in.
Quantity:
12/Cs.
144/Cs.
24/Cs.
384/Cs.
48/Cs.
Description
- Provides content visibility
- Closure (Green thermoset (phenolic)) provides the chemical compatibility and most temperature tolerant
- Foam backed PTFE liners provides resilience for tight seal with low moisture transmission rate
Analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Specifications
Specifications
Capacity | 1 oz., 30mL |
Capacity (Metric) | 30 mL |
Capacity (English) | 1 oz. |
Color | Clear |
Diameter (English) | 1.69 in. |
Material | Glass |
Quantity | 384/Cs. |
Closure Color | Green |
Height (Metric) | 43 mm |
Height (English) | 1.69 in. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction