Learn More
World Precision Instrument Quad 4-Axis Motorized Micromanipulator-SU-QUAD-R
SDP
Supplier:  World Precision Instrument SUQUADR
Encompass_Preferred
The QUAD motorized micromanipulator is easy to use and features four independent axes. Each axis has a 25 mm range of motion, a digital display of position and a control box. The compact, intuitive controller takes up minimal bench space, is fan free and is easy to use. Three axes provide the X, Y, Z-orthogonal motion typical of most motorized micromanipulators. The QUAD features a true fourth axis to move the electrode coaxially at exactly the desired angle of approach. The fourth axis has 30 mm of travel that significantly extends the range of travel for the system.
Catalog No. 50-253-8511
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.