Learn More
Electron Microscopy Sciences Quality Control Testing Standard Set A
Supplier:  Electron Microscopy Sciences 8015025
Encompass_Preferred
For Scanning Electron Microscopes with Energy Dispersive or Wavelength Dispersive X-ray systems to check the performance and ensure performance is within the manufactures specification.
Cobalt, compulsory for checking Energy Dispersive analyzers, and any other five selected standards*
Duplex Brass for checking the contrast efficiency of Back Scattered detectors and detecting two major copper/zinc phases of 0.1 atomic number difference.
A Faraday Cage for measuring the beam current at the specimen plane is in the center.
S168T gold crystals on a carbon background for checking the performance of the SEM at high-resolution with a finer than a 10 micron probe.
S1930 silicon test specimen with squares of periodicity 9.9 micrometers lines about 1.9 micrometers is used for magnification calibration and assessing image distribution.
*Excludes SRM 481, SRM 482 and SRM 1872 On 25mm diameter block.
Catalog No. 50-282-12
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.