Learn More
Ted Pella Inc QUANTIFOIL 1.2UM 300M CU 100PK
SDP
Supplier:  Ted Pella Inc 658300CU100
Encompass
Quantifoil, Orthogonal Array of 1.2µm Diameter Holes - 1.3µm Separation, mounted on a 300M Cu grid, package of 100 grids
Catalog No. NC1811329
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.