Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Homogenizing Solution
SDP

Catalog No. p-7180497
Click to view available options
Quantity:

QuantiGene Homogenizing Solution is used to prepare tissue homogenates from fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues.

QuantiGene Homogenizing Solution is used to prepare tissue homogenates from fresh, frozen, or formalin-fixed paraffin-embedded (FFPE) tissues. Homogenization of tissues using QuantiGene Homogenizing Solution releases RNA for detection and quantitation with QuantiGene assays. The homogenizing solution is one of two components in the QuantiGene Sample Processing Kit for tissues, and is used to supplement this kit when extra volume is needed.

Specifications

Content And Storage

Store at room temperature.

Sample Type

FFPE Tissue

For Use With (Application)

Multi-Gene Expression

Product Line

QuantiGene™

Product Type

Cell Lysis Buffer

Quantity

20 mL

Shipping Condition

Blue Ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.