Learn More
CTL Scientific Supply Corp QUANTOFIX TOTAL IRON 100
SDP
Supplier:  CTL Scientific Supply Corp 91344
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Test sicks/new version/necessary reagent already added to test pad. Gradation 0 - 2 - 5 - 10- 25- 50 -100 mg/L Fe 2+/3+ 100 tests/box
Catalog No. NC0220277