Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quercetin, Tocris Bioscience™

Catalog No. 1125100 Shop All R&D Systems Products
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective PI 3-kinase inhibitor

Antitumor agent; induces apoptosis and inhibits synthesis of heat shock proteins. Inhibits many enzyme systems including tyrosine protein kinase, phospholipase A2, phosphodiesterases, mitochondrial ATPase, PI 3-kinase and protein kinase C. Can also activate Ca2+ and K+ channels and behaves as an agonist at estrogen (GPER) receptors. Blocks cellular entry by influenza A virus via an interaction with hemaglutinin. Inhibits SARS-CoV 3CLpro (IC50 = 73 μM).

Chemical Identifiers

CAS 117-39-5
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.238
InChI Key REFJWTPEDVJJIY-UHFFFAOYSA-N
Synonym quercetin, sophoretin, meletin, xanthaurine, quercetine, quercetol, quercitin, quertine, 3,3',4',5,7-pentahydroxyflavone, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one
PubChem CID 5280343
ChEBI CHEBI:16243
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=C(C(=O)C3=C(C=C(C=C3O2)O)O)O)O)O

Specifications

Quantity 100 mg
Formula Weight 302.24
Percent Purity >98%
Chemical Name or Material Quercetin
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.