Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinhydrone, MP Biomedicals™

Catalog No. MP215648280 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Can be used to measure the hydrogen ion concentration (pH) of a solution in a chemical experiment. Quinhydrone, MP Biomedicals is a type of redox electrode that is an addition compound of 1M hydroquinone and 1M benzoquinone.

Quinhydrone/methanol treatment for the measurement of carrier lifetime in crystalline silicon substrates has been reported. Surface passivation of silicon substrates by quinhydrone/ethanol treatment has been investigated.

Chemical Identifiers

CAS

106-34-3

Molecular Formula

C12H10O4

Molecular Weight (g/mol)

218.21

MDL Number

MFCD00010310

InChI Key

BDJXVNRFAQSMAA-UHFFFAOYSA-N

Synonym

quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1

PubChem CID

7801

ChEBI

CHEBI:26491

IUPAC Name

benzene-1,4-diol; cyclohexa-2,5-diene-1,4-dione

SMILES

OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1

Specifications

Melting Point

167°C to 172°C (literature)

Quantity

100g

Solubility Information

Soluble in 90% methanol (2% v/v, clear, orange-brown solution); slightly soluble in cold water, hot water, ammonia, alcohol, ether; insoluble in petroleum ether.

Formula Weight

218.20

Physical Form

Dark Green Powder

Recommended Storage

Store at Room Temperature (15–30°C).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.