Learn More
Medchemexpress LLC HY-B1752A 5mg Medchemexpress, Quinpirole Hydrochloride CAS:85798-08-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1752A5MG
Encompass_Preferred
Medchemexpress, HY-B1752A 5mg Quinpirole HCl CAS:85798-08-9 Quinpirole Hydrochloride ((-)-LY 171555) is a high-affinity agonist of dopamine D2/D3 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0162
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.