Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Quinpirole hydrochloride, Tocris Bioscience™
GSA_VA

Catalog No. p-4783078 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective D2-like agonist

Selective dopamine D2 receptor agonist (Ki values are 4.8, ∽24, ∽30 and 1900 nM at D2, D3, D4 and D1 receptors respectively).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 85798-08-9
Molecular Formula C13H22ClN3
Molecular Weight (g/mol) 255.79
InChI Key HJHVRVJTYPKTHX-HTMVYDOJSA-N
Synonym --quinpirole hydrochloride, quinpirole hydrochloride, quinpirole hcl, unii-t6i2w5v2k1, quinpirole hydrochloride usan, dsstox_cid_25519, dsstox_rid_80926, dsstox_gsid_45519, --4ar,8ar-4,4a,5,6,7,8,8a,9-octahydro-5-propyl-1h-pyrazolo 3,4-g quinoline monohydrochloride
PubChem CID 55397
IUPAC Name (4aR,8aR)-5-propyl-1,4,4a,6,7,8,8a,9-octahydropyrazolo[3,4-g]quinoline;hydrochloride
SMILES CCCN1CCCC2C1CC3=C(C2)NN=C3.Cl

Specifications

Quantity 10 mg
Formula Weight 255.79
Percent Purity >98%
Chemical Name or Material (-)-Quinpirole hydrochloride
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.