Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-2,2′-Dimethyl-1,1′-binaphthyl 98.0+%, TCI America™
SDP

Catalog No. p-7127529 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 19634-89-0
Molecular Formula C22H18
Molecular Weight (g/mol) 282.386
MDL Number MFCD00048075
InChI Key KDHFKMDVFWYSPT-UHFFFAOYSA-N
PubChem CID 141748
IUPAC Name 2-methyl-1-(2-methylnaphthalen-1-yl)naphthalene
SMILES CC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)C

Specifications

Melting Point 84°C
Color White
Quantity 1 g
Formula Weight 282.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-2,2′-Dimethyl-1,1′-binaphthyl
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.