Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-2-Acetoxypropionyl Chloride 98.0+%, TCI America™
SDP

Catalog No. p-7118212 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 53636-19-4
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.558
MDL Number MFCD27925055
InChI Key ALHZEIINTQJLOT-GSVOUGTGSA-N
Synonym (+)-O-Acetyl-D-lactoyl Chloride
PubChem CID 15557611
IUPAC Name [(2R)-1-chloro-1-oxopropan-2-yl] acetate
SMILES CC(C(=O)Cl)OC(=O)C

Specifications

Color Yellow
Boiling Point 53°C
Quantity 1 g
UN Number 3265
Formula Weight 150.56
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Acetoxypropionyl Chloride
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.