Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-alpha-Lipoic Acid 98.0+%, TCI America™
SDP

Catalog No. p-7131868 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g
100 g

Chemical Identifiers

CAS 1200-22-2
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD01631142
InChI Key AGBQKNBQESQNJD-SSDOTTSWSA-N
Synonym (R)-Thioctic Acid, (R)-1,2-Dithiolane-3-valeric Acid
PubChem CID 6112
ChEBI CHEBI:30314
IUPAC Name 5-[(3R)-1,2-dithiolan-3-yl]pentanoic acid
SMILES OC(=O)CCCC[C@@H]1CCSS1

Specifications

Melting Point 49°C
Color Yellow
Quantity 5 g
Formula Weight 206.32
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-alpha-Lipoic Acid
EINECSNumber (5)-0535
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.