Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Blasticidin S HCl

Catalog No. 550225 Shop All R&D Systems Products
Click to view available options
:
25mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antibiotic; selection reagent for bis, bsr and BSD transformed cells

Blasticidin S HCl is an antibiotic. Selection reagent for bis, bsr and BSD transformed cells.

Specifications

Chemical Name or Material Blasticidin S HCl
CAS 3513-03-9
Synonym (S)-4-[[3-Amino-5-[(aminoiminomethyl)methylamino]-1-oxopentyl]amino]-1-(4-amino-2-oxo-1(2H)-pyrimidinyl)-1,2,3,4-tetradeoxy-β-D-erythro-hex-2-enopyranuronic acid hydrochloride
Quantity 25 mg
Solubility Soluble to 10mM in water
Molecular Formula C17H26N8O5·HCl
Formula Weight 458.9
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.