Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Adenosine A2aR (NP_000666) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
10ug
Description
The Human Adenosine A2a R (NP_000666) VersaClone cDNA clone contains the complete ORF for Adenosine A2aR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Adenosine A2aR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Gene | hADORA2A |
Accession Number | NP_000666 |
Concentration | 10 μg at 0.2 μg/μL |
Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
Insert Type | 1252bp |
Gene Alias | A2aR, adenosine A2 receptor, Adenosine A2a R, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, ADORA2A, hA2aR, RDC8 |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Human |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction