Learn More
R&D Systems™ Human Connexin 36/GJD2 (NP_065711) VersaClone cDNA
Shop All R&D Systems ProductsOrder Info
Ships at ambient temperature.
Specifications
Product Type
cDNA
Quantity
10μg
Species
Human, Mouse, Rat
Description
Gene ID (Entrez): 57369, 14617, 50564
Gene
hGJD2
Accession Number
NP_065711
Concentration
10μg at 0.2μg/μL
Formulation
cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type
979bp
Storage Requirements
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
For Research Use Only; Not for Use in Humans
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.