Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human P2Y12/P2RY12 (NP_073625) VersaClone cDNA
Shop All R&D Systems Products
Click to view available options
:
10ug
Description
The Human P2Y12/P2RY12 (NP_073625) VersaClone cDNA clone contains the complete ORF for P2Y12/P2RY12, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The P2Y12/P2RY12 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Gene | hP2RY12 |
Accession Number | NP_073625 |
Concentration | 10 μg at 0.2 μg/μL |
Formulation | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
Insert Type | 1041bp |
Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Species | Human |
For Reasearch Use Only; Not for Use in Humans
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction