Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human PTGER4/EP4 (NP_000949) VersaClone cDNA

Catalog No. RDC0176 Shop All R&D Systems Products
Click to view available options
:
10ug
This item is not returnable. View return policy
This item is not returnable. View return policy

Prostaglandin E2 Receptor 4, Subtype EP4

The Human PTGER4/EP4 (NP_000949) VersaClone cDNA clone contains the complete ORF for PTGER4/EP4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PTGER4/EP4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

Specifications

Gene hPTGER4
Accession Number NP_000949
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10mM Tris-Cl, pH 8.5
Insert Type 1480bp
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human

For Reasearch Use Only; Not for Use in Humans

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.