Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human TGF-beta RII (NP_003233) VersaClone cDNA

Catalog No. RDC1270 Shop All R&D Systems Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

TGF-beta receptor type IIB; TGF-beta receptor type-2; TGF-beta type II receptor; TGFbeta-RII; TGFR-2; transforming growth factor beta receptor type IIC

The Human TGF-beta RII (NP_003233) VersaClone cDNA clone contains the complete ORF for TGF-beta RII, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TGF-beta RII insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature

Specifications

Product Type

cDNA

Quantity

10μg

Species

Human

Gene

hTGFBR2

Accession Number

NP_003233

Concentration

10μg at 0.2μg/μL

Formulation

cDNA is provided in 10mM Tris-Cl, pH 8.5

Insert Type

1717bp

Storage Requirements

Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

Gene ID (Entrez)

7048, 21813

Host Species

Human, Mouse

For Reasearch Use Only; Not for Use in Humans.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.