Learn More
R&D Systems™ Me4BodipyFL-Ahx3Leu3VS (Proteasome Activity Probe)
A cell-permeable fluorescent proteasome activity probe for accurate profiling of proteasomal activity in cell lysates, intact cells, and murine and human patient-derived material, with high sensitivity
Supplier: R&D Systems™ I190050
Description
- For cell lysate labeling and general flow cytometry experiments
- Fluorescence can be monitored using excitation 515nm and emission 519nm wavelengths
- Purity >95% by HPLC
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
50 μg | |
C54H89BF2N8O8S Ubiquitinase Inhibitor |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.