Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NS 8593 Hydrochloride
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
NS 8593 hydrochloride is a selective KCa2 (SK) channel negative modulator; inhibits SK channel currents (Kd values are 0.42, 0.6 and 0.73 μM for SK1, SK2 and SK3 respectively at 0.5 μM Ca2+). Exhibits selectivity for SK channels over KCa1.1 (BK), KCa3.1 (IK), Kv, Nav and Cav channels. Inhibits afterhyperpolarization in hippocampal slices.
Specifications
Specifications
Chemical Name or Material | NS 8593 hydrochloride |
CAS | 875755-24-1 |
Synonym | N-[(1R)-1,2,3,4-Tetrahydro-1-naphthalenyl]-1H-benzimidazol-2-amine hydrochloride |
Quantity | 10 mg |
Solubility | Soluble to 100mM in DMSO and to 20mM in ethanol |
Molecular Formula | C17H17N3·HCl |
Formula Weight | 299.8 |
Physical Form | Solid |
Product Type | Bioactive Small Molecule |
Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction