Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat V1a Vasopressin R/AVPR1A (NP_444178) VersaClone cDNA

Catalog No. RDC0747 Shop All R&D Systems Products
Click to view available options
:
This item is not returnable. View return policy
This item is not returnable. View return policy

Arginine Vasopressin Receptor 1A

The Rat V1a Vasopressin R/AVPR1A (NP_444178) VersaClone cDNA clone contains the complete ORF for V1a Vasopressin R/AVPR1A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The V1a Vasopressin R/AVPR1A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.

Order Info

Ships at ambient temperature.

Specifications

Gene

rAVPR1A

Accession Number

NP_444178

Concentration

10 μg at 0.2 μg/μL

Formulation

cDNA is provided in 10mM Tris-Cl, pH 8.5

Insert Type

1287bp

Content And Storage

Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.

Species

Rat

For Reasearch Use Only; Not for Use in Humans

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.