Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Glycidyl Trityl Ether 98.0+%, TCI America™
SDP

Catalog No. p-7130228 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 65291-30-7
Molecular Formula C22H20O2
Molecular Weight (g/mol) 316.4
MDL Number MFCD00273368
InChI Key XFSXUCMYFWZRAF-OAQYLSRUSA-N
Synonym (R)-Glycidyl Triphenylmethyl Ether, (R)-2-(Triphenylmethoxymethyl)oxirane
PubChem CID 2734442
IUPAC Name (2R)-2-(trityloxymethyl)oxirane
SMILES C1C(O1)COC(C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 102°C
Color White
Quantity 5 g
Formula Weight 316.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Glycidyl Trityl Ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.