Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-2,2′-Isopropylidenebis(4-phenyl-2-oxazoline) 96.0+%, TCI America™
SDP

Catalog No. p-7127519 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
250 mg
1 g
5 g

Chemical Identifiers

CAS 150529-93-4
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.42
MDL Number MFCD00192244
InChI Key JTNVCJCSECAMLD-UHFFFAOYNA-N
Synonym +-2,2'-isopropylidenebis 4r-4-phenyl-2-oxazoline, ph-box, r,r, unii-cg7p67w7w0, r,r-2,2'-isopropylidenebis 4-phenyl-2-oxazoline, r,r-2,2'-dimethylmethylene bis 4-phenyl-2-oxazoline, 4r,4'r-2,2'-propane-2,2-diyl bis 4-phenyl-4,5-dihydrooxazole, oxazole, 2,2'-1-methylethylidene bis 4,5-dihydro-4-phenyl-, 4r,4'r, r,r-2,2-bis 4-phenyl-2-oxazolin-2-yl propane, r,r-2,2-dimethylmethylene bis 4-phenyl-2-oxazoline
PubChem CID 676390
IUPAC Name 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 334.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R,R)-2,2′-Isopropylidenebis(4-phenyl-2-oxazoline)
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.