Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R,R)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine 98.0+%, TCI America™
SDP

Catalog No. p-7128593 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 415918-91-1
Molecular Formula C37H32Cl2NO2P
Molecular Weight (g/mol) 624.542
MDL Number MFCD08561138
InChI Key PQTVIIOJNJVQOL-JUJAXGASSA-N
Synonym (11bR)-N,N-Bis[(1R)-1-phenylethyl]dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine
PubChem CID 72376409
SMILES CC(C1=CC=CC=C1)N(C(C)C2=CC=CC=C2)P3OC4=C(C5=CC=CC=C5C=C4)C6=C(O3)C=CC7=CC=CC=C76.C(Cl)Cl

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 539.61
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R,R)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.