Learn More
Indigo Biosciences Rabbit Peroxisome Proliferator-Activated Receptor delta (rPPARd, NR1C2) All-inclusive Cell-Based Luciferase Reporter Assay Kit, 96-well assay plate format
Supplier: Indigo Biosciences RB00121
Eliminate weeks of cell culture while achieving superior sensitivity with reproducible results from this all-inclusive cell-based luciferase reporter assay. INDIGO’s Rabbit Peroxisome Proliferator-Activated Receptor delta (rPPARd, NR1C2) reporter assay allows you to quickly screen test samples to quantify functional activity, either agonist or antagonist, they may exert against Rabbit PPARd. Kit includes cryopreserved reporter cells engineered to provide constitutive expression of the receptor, optimized media for use during cell thawing and in diluting test samples, reference agonist, Luciferase Detection Reagent, cell culture-ready assay plate, and detailed assay protocol. INDIGO’s unique Cryomite™ process provides exceptional cell viability post-thaw and INDIGO’s strict QA process means consistent results lot to lot (product quality data available upon request).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.