Learn More
Medchemexpress LLC HY-15394 10mg Medchemexpress, (Rac)-Rotigotine (hydrochloride) CAS:102120-99-0 Purity:>98%
Supplier: Medchemexpress LLC HY1539410MG
Medchemexpress, HY-15394 10mg (Rac)-Rotigotine (hydrochloride) CAS:102120-99-0 (Rac)-Rotigotine hydrochloride is a racemate of Rotigotine. Rotigotine is a full agonist of dopamine receptor, a partial agonist of the 5-HT1A receptor, and an antagonist of the α2B-adrenergic receptor, with Kis of 0.71 nM, 4-15 nM, and 83 nM for the dopamine D3 receptor and D2, D5, D4 receptors, and dopamine D1 receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.