Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Racked Micro Tips, Sterile



These Sterile Racked Micro Tips are disposable tips intended for use with most brands of pipettors.
Supplier: United Scientific™ UNP10148
Description
- The highly smooth, hydrophobic surface provides low liquid retention and increased accuracy and repeatability.
- Inner radial rings enable an airtight grip on the tip cone.
- Manufactured in a precision molding facility from 100% virgin polypropylene.
- Free of DNase, RNase and pyrogen.
Specifications
Racked Micro Tips, Sterile, 2 to 200 μL, Natural | |
Non-filtered | |
Natural | |
Autoclavable | |
USP Class VI certified | |
Steam | |
Rack, 10 x 96 | |
10 Racks of 96 | |
Micro Tips | |
No |
200 μL | |
Sterile | |
Blue | |
UNP10148 | |
Pipetting | |
Universal | |
Standard | |
960 Pk. | |
Reusable | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction