Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raclopride, Tocris Bioscience™

Catalog No. p-4784389 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective D2/D3 antagonist

Selective and potent dopamine D2/D3 receptor antagonist (Ki values are 1.8, 3.5, 2400 and 18000 nM for D2, D3, D4 and D1 receptors respectively). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 84225-95-6
Molecular Formula C15H20Cl2N2O3
Molecular Weight (g/mol) 347.236
InChI Key WAOQONBSWFLFPE-VIFPVBQESA-N
Synonym raclopride, s-3,5-dichloro-n-1-ethylpyrrolidin-2-yl methyl-2-hydroxy-6-methoxybenzamide, 3h raclopride, 3h-raclopride, unii-430k3soz7g, chembl8809, raclopridum latin, racloprida spanish, raclopride inn:ban, dsstox_cid_25687
PubChem CID 3033769
IUPAC Name 3,5-dichloro-N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-hydroxy-6-methoxybenzamide
SMILES CCN1CCCC1CNC(=O)C2=C(C(=CC(=C2OC)Cl)Cl)O

Specifications

Quantity 10 mg
Formula Weight 347.24
Percent Purity 98%
Chemical Name or Material Raclopride
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.