Learn More
Thermo Scientific™ RadHalo FM Fixed Spectroscopic Area Monitors

Monitor radiation with high sensitivity and accuracy
Supplier: Thermo Scientific™ RH FMR
Description
The Thermo Scientific RadHalo FM Spectroscopic Area Monitor is designed for permanent and constant area monitoring. Able to withstand nearly all environmental conditions and emergency conditions. Battery backup and multiple reachback methods insure full operation even in emergency situations. Simple layout allows field service support and onsite calibrations. Antenna and satellite extension kits for long distance communication capability especially when RadHalo has no view of the sky.- Rapid dose rate response
- Immediate accurate identification
- Autonomous operation anywhere
- Durable design
- Secure transmissions
- Detector Fusion Capable
Specifications
Spectroscopic Detector: 3 x 3 in. NaI | |
RadHalo FM-R | |
With Radio |
None | |
Wi-Fi, 2.4 GHz long range radio and Cellular reachback Capabilty |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.