Learn More
MilliporeSigma™ Supelco™ radiello™ Cartridge Adsorbents For Sampling Hydrogen Sulfide (H2S), Matrix microporous PE coated with zinc acetate
For Sampling Hydrogen Sulfide (H2S)
Supplier:  MilliporeSigma™ Supelco™ RAD170
Catalog No. 11-100-9314
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Several different radiello™ cartridge adsorbents are available. Each of which are specific for sampling different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm D. Each cartridge is designed for one time use with the exception of radiello™ thermal desorption (TD) cartridge adsorbents. Each cartridge arrives in a sealed glass or plastic tube wrapped in a transparent thermally sealed polyethylene (PE) bag. The same sealed tube can be used to store the cartridge after sampling prior to desorption/analysis. A sufficient number of adhesive barcode labels are included with every pack of cartridges for easy tracking of sampling date and time. Note: To conduct air sampling using the radiello™ system, radiello™ diffusive bodies and triangular support plates must be ordered (separately) in addition to cartridge adsorbents. radiello™ Starter Kits (RADxxxS) include all the necessary parts to complete a sampling.
Specifications
Specifications
radiello
20/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
radiello is a trademark of Institi Clinici Scientifici Maugeri