Learn More
Cell Biolabs Inc RAF-1 RBD AGAROSE BEADS
Supplier:  Cell Biolabs Inc STA410
Encompass_Preferred
Raf-1 RBD Agarose Beads selectively pull down the active form of Ras. Beads are colored to allow for a visual check. These are the same beads supplied with our Ras Activation Assays. 400 ug.
Catalog No. NC0736415
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.