Learn More
Hand2Mind Rainbow Fraction™ Liquid Measuring Cups

- These colorful, food-safe, dishwasher-safe, liquid measuring cups provide a hands-on way for children to learn measurement concepts such as volume, capacity, and fractions while cooking or engaging in imaginative play.
- Plus, the nesting design means they can double as a stacking toy.
Specifications
Product Type
Fractions, Decimals, Percents
Grade
K to 12
Class Size
1 Student
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.