Learn More
Chemglass Life Sciences Rapid Purge Valve, Adjustable, Complete
SDP
Supplier:  Chemglass Life Sciences CG453501
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Valve, Check. Similar to CG-4539, but having serrated hose connections on either end to accept flexible tubing. Tubing O.D. is 10mm while the overall length is approx. 100mm. Hose connection O.D. is 9mm at the middle ring.
Catalog No. 50-873-465