Learn More
Innovative Research Inc Rat ADAMS 10 Kit
Supplier: Innovative Research Inc IRTADAM10KT
Rat A Disintegrin And Metalloprotease 10 ELISA Kit - This kit from Innovative Research is intended for the quantitative determination of Rat A Disintegrin And Metalloprotease 10 in biofluid samples, such as serum, plasma, tissue homogenates and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . ADAM10 present in samples reacts with an anti-Rat ADAM10 capture antibody coated and dried on a microtiter plate. After appropriate washing steps, an anti-ADAM10 detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is directly proportional to the concentration of active ADAM10 in the samples. A standard calibration curve is prepared along with the samples to be measured using dilutions of Rat ADAM10. For RUO.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.