Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raw 264.7 (mouse macrophage, Abelson murine leukemia virus transformed) Nuclear extract lysate, Denatured; Abnova

Catalog No. 89014481 Shop All Abnova Corporation Products
Click to view available options
:

Western Blotting

Specifications

For Use With (Application)

Western Blot

Tissue

Macrophage

Description

Nuclear extract cell lysate (denatured)

Lysis Buffer

Buffer A: 10mM HEPES pH 7.9, 1.5mM MgCl2, 10mM KCl, 0.5 mM DTT. Buffer C: 20mM HEPES pH 7.9, 25%(v/v) Glycerol , 0.42M NaCl , 1.5mM MgCl2, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF. Buffer D : 20mM HEPES pH 7.9, 20%(v/v) glycerol, 50mM KCl, 0.2 mM EDTA, 0.5 mM DTT & 0.5 mM PMSF.

Preparation Method

Nuclear extract was prepared by using a modified protocol of Dignam et al. Cells were Harvested and homogenized in Buffer A, and then centrifugated at 25,000 g for 20 minutes to remove cytoplasm and pellet the nuclei. The pellet was re-suspended in Buffer C, and then the suspensions were centrifuged to collect nuclear extract. The supernatant was dialyzed against Buffer D. The dialysate was then centrifuged, divided into aliquots, and stored at -80°C. The protein concentration was determined by the method of Bradford (Bio-Rad protein assay, microplate standard assay). The lysate was adjusted to 2.5 mg/ml, and then mixed with 5X Sample Buffer to become final 2 mg/ml in 1X Sample Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly.

Quality Control Testing

12.5% SDS-PAGE Stained with Coomassie Blue.

Quantity

50 μg

Storage Buffer

In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue).

Host Species

Mouse

Content And Storage

Store at -80°C. Aliquot to avoid repeated freezing and thawing.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.