Learn More
RB705 Mouse Anti-Human CD45, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757307
Description
CD45 Monoclonal antibody specifically detects CD45 in Human samples. It is validated for Flow cytometrySpecifications
CD45 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
HI30 | |
RB705 | |
PTPRC; LCA; L-CA; Leukocyte Common Antigen; T200; GP180; LY5 | |
Human Peripheral Blood Leucocytes | |
50 μg | |
5788 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.