Learn More
RB780 Mouse Anti-Insulin, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570442
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
Insulin | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
T56-706 | |
RB780 | |
INS; IDDM; ILPR; IRDN; IDDM1; IDDM2; MODY10; Ins2 | |
Mature human insulin (A- and B-chain) Recombinant Protein | |
25 μg | |
3630, 16334, 24506 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.