Learn More
RB780 Rat Anti-Mouse CD301b (Mgl2), BD Horizon™
Rat Monoclonal Antibody
$194.00 - $422.00
Specifications
Antigen | CD301b (Mgl2) |
---|---|
Clone | URA-1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD301b (Mgl2) Monoclonal antibody specifically detects CD301b (Mgl2) in Mouse samples. It is validated for Flow cytometrySpecifications
CD301b (Mgl2) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
216864 | |
IgG2a λ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
URA-1 | |
Flow Cytometry | |
RB780 | |
Rat | |
Mouse | |
CD301b; Mgl2; macrophage galactose-type C-type lectin 2 | |
Recombinant Mouse MGL2 Protein | |
Affinity Chromatography |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.