Learn More
RB780 Rat Anti-TOX, BD Horizon™
Rat Monoclonal Antibody
$194.00 - $422.00
Specifications
Antigen | TOX |
---|---|
Clone | NAN448B |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
TOX Monoclonal antibody specifically detects TOX in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
TOX | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
9760, 252838 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
NAN448B | |
Flow Cytometry | |
RB780 | |
Rat | |
Human, Mouse | |
TOX; TOX 1; TOX1 | |
Recombinant Human TOX Protein | |
Affinity Chromatography |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.